Anti-UHRF2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7228
BWRLBS7228EA 341 EUR
BWRLBS7228
Anti-UHRF2 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human UHRF2.

UHRF2 pAb detects endogenous levels of UHRF2 protein.

Type: Primary
Antigen: UHRF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR