Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Di-Methyl K27]

Lieferant: Bioworld Technology
BS7236
BWRLBS7236EA 379 EUR
BWRLBS7236
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Di-Methyl K27]
Antikörper
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, di methylated at K27.

Histone H3 (Di-Methyl K27) pAb detects endogenous levels of histone H3 only when di-methylated at Lys27.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: Di-Methyl K27
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat; Other (Wide Range)
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR