Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Mono-Methyl K36]

Lieferant: Bioworld Technology
BS7238
BWRLBS7238EA 379 EUR
BWRLBS7238
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Mono-Methyl K36]
Antikörper
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, mono methylated at K36.

Histone H3 (Mono-Methyl K36) pAb detects endogenous levels of histone H3 only when mono-methylated at Lys36.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: Mono-Methyl K36
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat; Other (Wide Range)
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR