Anti-SR-3A Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7250
BWRLBS7250EA 341 EUR
BWRLBS7250
Anti-SR-3A Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human SR-3A.

SR-3A pAb detects endogenous levels of SR-3A protein.

Type: Primary
Antigen: SR-3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR