Anti-TXA synthase Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7276
BWRLBS7276EA 341 EUR
BWRLBS7276
Anti-TXA synthase Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human TXA synthase.

TXA synthase pAb detects endogenous levels of TXA synthase protein.

Type: Primary
Antigen: TXA synthase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR