Anti-DLD Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7282
BWRLBS7282EA 341 EUR
BWRLBS7282
Anti-DLD Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human DLD.

DLD pAb detects endogenous levels of DLD protein.

Type: Primary
Antigen: DLD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR