Anti-DLC1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7318
BWRLBS7318EA 341 EUR
BWRLBS7318
Anti-DLC1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human DLC1.

DLC1 pAb detects endogenous levels of DLC1 protein.

Type: Primary
Antigen: DLC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR