Anti-U2AF2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7326
BWRLBS7326EA 341 EUR
BWRLBS7326
Anti-U2AF2 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human U2AF2.

U2AF2 pAb detects endogenous levels of U2AF2 protein.

Type: Primary
Antigen: U2AF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR