Anti-SERPINH1 Rabbit Polyclonal Antibody
Lieferant: Bioworld Technology
BS7370
BWRLBS7370EA 341 EUR
BWRLBS7370
Anti-SERPINH1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human SERPINH1.
SERPINH1 pAb detects endogenous levels of SERPINH1 protein.
Type: Primary
Antigen: SERPINH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat