Anti-GPC3 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7410
BWRLBS7410EA 341 EUR
BWRLBS7410
Anti-GPC3 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human GPC3.

  • GPC3 pAb detects endogenous levels of GPC3 protein.

Type: Primary
Antigen: GPC3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR