Anti-NLK Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7516
BWRLBS7516EA 341 EUR
BWRLBS7516
Anti-NLK Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human NLK.

NLK pAb detects endogenous levels of NLK protein.

Type: Primary
Antigen: NLK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR