Anti-HIF1AN Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7590
BWRLBS7590EA 341 EUR
BWRLBS7590
Anti-HIF1AN Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human HIF1AN.

HIF1AN pAb detects endogenous levels of HIF1AN protein.

Type: Primary
Antigen: HIF1AN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR