Anti-GLA Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7679
BWRLBS7679EA 341 EUR
BWRLBS7679
Anti-GLA Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human GLA.

GLA pAb detects endogenous levels of GLA protein.

Type: Primary
Antigen: GLA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR