Anti-AP2B1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7713
BWRLBS7713EA 341 EUR
BWRLBS7713
Anti-AP2B1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human AP2B1.

AP2B1 pAb detects endogenous levels of AP2B1 protein.

Type: Primary
Antigen: AP2B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR