Anti-GTF2F1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7719
BWRLBS7719EA 341 EUR
BWRLBS7719
Anti-GTF2F1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human GTF2F1.

GTF2F1 pAb detects endogenous levels of GTF2F1 protein.

Type: Primary
Antigen: GTF2F1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR