Anti-ACTR2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7823
BWRLBS7823EA 341 EUR
BWRLBS7823
Anti-ACTR2 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human ACTR2.

ACTR2 pAb detects endogenous levels of ACTR2 protein.

Type: Primary
Antigen: ACTR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR