Anti-GNA13 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7833
BWRLBS7833EA 341 EUR
BWRLBS7833
Anti-GNA13 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human GNA13.

GNA13 pAb detects endogenous levels of GNA13 protein.

Type: Primary
Antigen: GNA13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR