Anti-PEX5 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7857
BWRLBS7857EA 341 EUR
BWRLBS7857
Anti-PEX5 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human PEX5.

PEX5 pAb detects endogenous levels of PEX5 protein.

Type: Primary
Antigen: PEX5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR