Anti-BBOX1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7873
BWRLBS7873EA 341 EUR
BWRLBS7873
Anti-BBOX1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human BBOX1.

BBOX1 pAb detects endogenous levels of BBOX1 protein.

Type: Primary
Antigen: BBOX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR