Anti-SCNN1G Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7921
BWRLBS7921EA 341 EUR
BWRLBS7921
Anti-SCNN1G Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human SCNN1G.

SCNN1G pAb detects endogenous levels of SCNN1G protein.

Type: Primary
Antigen: SCNN1G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR