Anti-KAO Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7991
BWRLBS7991EA 341 EUR
BWRLBS7991
Anti-KAO Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human KAO.

KAO pAb detects endogenous levels of KAO protein.

Type: Primary
Antigen: KAO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR