Anti-c-Jun Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0004
BWRLMB0004EA 341 EUR
BWRLMB0004
Anti-c-Jun Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human c-Jun.

This antibody detects endogenous levels of JUN and does not cross-react with related proteins

Type: Primary
Antigen: c-Jun
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR