Anti-Tubulin beta Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0009
BWRLMB0009EA 341 EUR
BWRLMB0009
Anti-Tubulin beta Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human Tubulin beta.

This antibody detects endogenous levels of Tubulin beta and does not cross-react with related proteins

Type: Primary
Antigen: Tubulin beta
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse;Rat;Monkey;Goat;Hamster
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR