Anti-Cdk1/Cdc2 Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0019
BWRLMB0019EA 341 EUR
BWRLMB0019
Anti-Cdk1/Cdc2 Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human Cdk1/Cdc2.

This antibody detects endogenous levels of Cdk1/Cdc2 and does not cross-react with related proteins

Type: Primary
Antigen: Cdk1/Cdc2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR