Anti-PEG10 Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0036
BWRLMB0036EA 341 EUR
BWRLMB0036
Anti-PEG10 Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human PEG10.

This antibody detects endogenous levels of PEG10 and does not cross-react with related proteins

Type: Primary
Antigen: PEG10
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR