Anti-JMJD6 Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0100
BWRLMB0100EA 341 EUR
BWRLMB0100
Anti-JMJD6 Mouse Monoclonal Antibody
Antikörper
Synthetic peptide, corresponding to the N-terminus of Human JMJD6.

This antibody detects endogenous levels of JMJD6 and does not cross-react with related proteins

Type: Primary
Antigen: JMJD6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope: N-terminal
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR