Anti-PPP5C Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0112
BWRLMB0112EA 341 EUR
BWRLMB0112
Anti-PPP5C Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human PPP5C.

This antibody detects endogenous levels of PPP5C and does not cross-react with related proteins

Type: Primary
Antigen: PPP5C
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR