Anti-CDX2 Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0125
BWRLMB0125EA 341 EUR
BWRLMB0125
Anti-CDX2 Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human CDX2.

CDX2 mAb detects endogenous levels of CDX2 protein.

Type: Primary
Antigen: CDX2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Monkey;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR