Anti-Csk Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0131
BWRLMB0131EA 341 EUR
BWRLMB0131
Anti-Csk Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human Csk.

This antibody detects endogenous levels of Csk and does not cross-react with related proteins.

Type: Primary
Antigen: Csk
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR