Anti-GATA-6 Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0137
BWRLMB0137EA 341 EUR
BWRLMB0137
Anti-GATA-6 Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human GATA-6.

This antibody detects endogenous levels of GATA-6 and does not cross-react with related proteins

Type: Primary
Antigen: GATA-6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR