Anti-HSP90B Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0142
BWRLMB0142EA 341 EUR
BWRLMB0142
Anti-HSP90B Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human HSP90B.

This antibody detects endogenous levels of HSP90 and does not cross-react with related proteins

Type: Primary
Antigen: HSP90B
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse;Rat;Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR