Anti-LIN28A Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0149
BWRLMB0149EA 341 EUR
BWRLMB0149
Anti-LIN28A Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human LIN28A.

This antibody detects endogenous levels of LIN28A and does not cross-react with related proteins

Type: Primary
Antigen: LIN28A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR