Anti-MPI Mouse Monoclonal Antibody

Lieferant: Bioworld Technology
MB0150
BWRLMB0150EA 341 EUR
BWRLMB0150
Anti-MPI Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human MPI.

This antibody detects endogenous levels of Mannose Phosphate Isomerase and does not cross-react with related proteins

Type: Primary
Antigen: MPI
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR