Anti-NFkB-p105/p50 Mouse Monoclonal Antibody
Lieferant: Bioworld Technology
MB0155
BWRLMB0155EA 341 EUR
BWRLMB0155
Anti-NFkB-p105/p50 Mouse Monoclonal Antibody
Antikörper
Recombinant full length Human NFkB-p105.
This antibody detects endogenous levels of NFkB-p105/p50 and does not cross-react with related proteins
Type: Primary
Antigen: NFkB-p105/p50
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human