Anti-pp1ab Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
NCP0004
BWRLNCP0004EA 379 EUR
BWRLNCP0004
Anti-pp1ab Rabbit Polyclonal Antibody
Antikörper
pp1ab polyclonal antibody detects endogenous levels of pp1ab protein

  • For research and diagnosis

Type: Primary
Antigen: pp1ab
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 2019-nCoV
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR