Anti-ORF7a Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
NCP0011
BWRLNCP0011EA 379 EUR
BWRLNCP0011
Anti-ORF7a Rabbit Polyclonal Antibody
Antikörper
ORF7a polyclonal antibody detects endogenous levels of ORF7a protein

  • For research and diagnosis

Type: Primary
Antigen: ORF7a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 2019-nCoV
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR