Anti-ORF3a Rabbit Polyclonal Antibody
Lieferant: Bioworld Technology
NCP0019
BWRLNCP0019EA 379 EUR
BWRLNCP0019
Anti-ORF3a Rabbit Polyclonal Antibody
Antikörper
ORF3a polyclonal antibody detects endogenous levels of ORF3a protein
  • For research and diagnosis
Type: Primary
Antigen: ORF3a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 2019-nCoV