Anti-CD34 Mouse Monoclonal Antibody [clone: ICO-115]

Lieferant: Cayman Chemical
10004835-1
CAYM10004835-1EA 375 EUR
CAYM10004835-1
Anti-CD34 Mouse Monoclonal Antibody [clone: ICO-115]
Antikörper
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: ICO-115
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR