Anti-CD27 Mouse Monoclonal Antibody [clone: 203.6]
Lieferant: Cayman Chemical
10004837-1
CAYM10004837-1EA 446 EUR
CAYM10004837-1
Anti-CD27 Mouse Monoclonal Antibody [clone: 203.6]
Antikörper
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 203.6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human