Anti-CRTH2 Rabbit Polyclonal Antibody

Lieferant: Cayman Chemical
10004886-1
CAYM10004886-1EA 418 EUR
CAYM10004886-1
Anti-CRTH2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CRTH2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR