Anti-Nitrotyrosine Mouse Monoclonal Antibody (Biotin) [clone: 8C7.3]

Lieferant: Cayman Chemical
10006966-100
CAYM10006966-100EA 467 EUR
CAYM10006966-100
Anti-Nitrotyrosine Mouse Monoclonal Antibody (Biotin) [clone: 8C7.3]
Antikörper
Type: Primary
Antigen: Nitrotyrosine
Clonality: Monoclonal
Clone: 8C7.3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: species independent
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR