Anti-5-LO Rabbit Polyclonal Antibody
Lieferant: Cayman Chemical
10007820-1
CAYM10007820-1EA 645 EUR
CAYM10007820-1
Anti-5-LO Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: 5-LO
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, non-Human, Rat