Anti-GSK-3-beta Rabbit Polyclonal Antibody
Lieferant: Cayman Chemical
10009374-1
CAYM10009374-1EA 645 EUR
CAYM10009374-1
Anti-GSK-3-beta Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GSK-3-beta
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat