Anti-TPH Rabbit Polyclonal Antibody
Lieferant: Cayman Chemical
10009397-1
CAYM10009397-1EA 645 EUR
CAYM10009397-1
Anti-TPH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TPH
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rabbit