Anti-dynamin Sheep Polyclonal Antibody

Lieferant: Cayman Chemical
10009785-1
CAYM10009785-1EA 645 EUR
CAYM10009785-1
Anti-dynamin Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: dynamin
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR