Anti-PGIS Mouse Monoclonal Antibody [clone: 3C8]

Lieferant: Cayman Chemical
10247-1
CAYM10247-1EA 418 EUR
CAYM10247-1
Anti-PGIS Mouse Monoclonal Antibody [clone: 3C8]
Antikörper
Type: Primary
Antigen: PGIS
Clonality: Monoclonal
Clone: 3C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR