Anti-CysLT2 Rabbit Polyclonal Antibody
Lieferant: Cayman Chemical
120560-1
CAYM120560-1EA 418 EUR
CAYM120560-1
Anti-CysLT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CysLT2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human