Anti-LL-37 Rabbit Polyclonal Antibody
Lieferant: Cayman Chemical
15637-1
CAYM15637-1EA 418 EUR
CAYM15637-1
Anti-LL-37 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LL-37
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human