Anti-ARC Rabbit Polyclonal Antibody

Lieferant: Cayman Chemical
160737-1
CAYM160737-1EA 592 EUR
CAYM160737-1
Anti-ARC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARC
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR