TamiSolve® NxG-PS Solvents for Peptide Synthesis
Lieferant: CEM
Modern peptide synthesis agents can dramatically reduce waste generation, but challenges remain regarding the health and environmental impacts of commonly used solvents.
- Low toxicity and flammability
- An efficient solvent for solid-phase peptide synthesis
- No effect on amino acid solution life
- Available in large quantities
- Easy to implement
- High purity
TamiSolve is just as effective as DMF or NMP with significantly less flammability and toxicity, so it can be routinely used as a cleaning solvent for Liberty systems as an alternative to DMF or NMP. Additionally, it can often be used as an effective reaction solvent to completely replace DMF and NMP.
TamiSolve was used as both a cleaning solvent and a reaction solvent.